missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer™ Glass Stir Rods with Rubber Policeman

Soft borosilicate glass with rounded ends and removable rubber policeman.

Brand:  Cole Parmer™ 06369-54

Product Code. 11725848

  • £30.20 / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Borosilicate glass rod.
  • Rounded ends.
  • Pre-assembled.
  • Removable rubber policeman.
  • Glass is chemically resistant and can withstand temperatures up to 205°C without rubber policeman.
  • Autoclavable.
Specifications

Specifications

Autoclavable
8 in.
Glass, Rubber
Rubber policeman
Up to 205°C without rubber policeman
12/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers