Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand:  Hirschmann™ 8900393
Additional Details : Weight : 0.60300kg
 View more versions of this product
Product Code. 10678971
  • £19.55 / Each
Estimated Shipment: 23-09-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Specifications
Specifications
Hollow Stopper
Glass
NS 45/40
1/Ea.
For tight sealing of standard ground vessels