Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand:  Hirschmann™ 8900352
 View more versions of this product
Product Code. 30090416
  • £44.15 / Pack of 10
Explore more special offers
Description
Description
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Specifications
Specifications
For tight sealing of standard ground vessels
NS 7/16
Hollow Stopper
1/Ea.
Glass