missing translation for 'onlineSavingsMsg'
Learn More

S Murray™ Glass Syringes

Use these syringes for a variety of laboratory applications. S Murray™ Glass Syringes are manufactured from precision ground, heat resistant glass.

Brand:  S Murray™ G811/32

Code : 02

Additional Details : Weight : 0.24300kg

Product Code. 12359249

  • £53.25 / Each
In Stock
to see stock.

Description

Description

  • Gas
  • 1ml graduations
  • Capacity: 100mL x 1mL
Specifications

Specifications

Samco Syringe
G811/32
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers