Learn More
BUCHI Glass Woulff Bottle Flask
Shop All Buchi Products
Click to view available options
Volume (Metric):
800 mL
Unit Size:
Each
Description
- Use a Woulff bottle to aid in trapping particles and droplets before they get to the vacuum source
- Benefit from pressure equalization between a vacuum pump and Rotavapor
Specifications
Specifications
| Material | Glass |
| Type | Woulff Flask |
| Material (Coating) | Plastic |
| Quantity | 1 Ea. |
| Volume (Metric) | 800 mL |