missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Glassware Caps for RapidVap™ Evaporation Systems
Shop All Labconco Corporation Products
Click to view available options
Material:
PTFE
Unit Size:
Pack of 8
Description
For use with RapidVap borosilicate glassware tubes. Placing Caps on Tubes helps prevent cross contamination and moisture from condensing when using low boiling point solvents. The use of caps increases overall evaporation time 10 to 80 percent. Caps are available in polyethylene or PTFE. Eight polyethylene glassware caps are included with each RapidVap N2 Evaporation System.
Specifications
Specifications
| Material | PTFE |
| Type | Glassware Cap |
| For Use With (Equipment) | 600mL glass tube of RapidVap™ evaporation system |
| Volume (Metric) | 600 mL |
| Weight (English) | 0.5 lb. |
| Weight (Metric) | 0.2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction