Learn More
Heidolph™ Glassware Set - Vertical Condenser with Valve (Reflux Distillation)
Vertical condenser, centerpiece with valve for reflux distillation
Brand:  Heidolph™ 513-00600-00
 View more versions of this product
Product Code. 10055741
  • £2425.00 / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glassware sets with a transparent plastic coating are suitable for a temperature range up to max. 80™C
  • Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
  • Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
Specifications
Specifications
Glass
Rotary Evaporators
None
Glassware Set
Glassware set G6 - Vertical condenser centerpiece with valve (reflux distillation), 1000 mL evaporating flask and 1000 mL receiving flask
1000 mL