missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glassy carbon plate, 1mm (0.04in) thick, type 1, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Packaging | Tube |
Quantity | 50 x 50 mm |
Solubility Information | Insoluble. |
Chemical Name or Material | Glassy carbon plate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'