missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glibenclamide, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
ATP-dependent K+ channel (Kir6, KATP) and CFTR Cl- channel blocker. Inhibits Kir6 currents in the pancreas, causing an increase in intracellular Ca2+ and insulin secretion. Inhibits recombinant CFTR Cl- channels with an IC50 of 20 μM.
Specifications
CAS | 10238-21-8 |
Molecular Formula | C23H28ClN3O5S |
Quantity | 100 mg |
Synonym | glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil |
InChI Key | ZNNLBTZKUZBEKO-UHFFFAOYSA-N |
SMILES | COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3 |
IUPAC Name | 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide |
Molecular Weight (g/mol) | 494.003 |
PubChem CID | 3488 |
ChEBI | CHEBI:5441 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'