missing translation for 'onlineSavingsMsg'
Learn More
Marigold™ Gloves Product Code.: 12951085

Marigold™ Gloves

Product Code. 12951085
Size: 6.5/Small
Click to view available options
Size: 6.5/Small
This item is not returnable. View return policy

Product Code. 12951085

Brand: Marigold™ S600PFM

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
A highly sensitive and versatile glove. Made of natural rubber combining high elasticity with maximum strength to give the wearer unimpeded hand movement. The microroughened surface ensures maximum grip and the beaded cuff a good fit. Ambidextrous. Natural color.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE0321,
Color Clear
Disposable Single-use
Product Type Gloves
Size Medium
Thickness (Metric) 0.2 mm
For Use With (Application) Ideal for use in laboratories with short-term chemical contact, light duty maintenance, painting, printing, pharmaceutical production and applications requiring good dexterity.
Quantity 100 Pk.
Length (Metric) 240 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Marigold™ Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.