missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Sol-Knit™ 39-122 Series Nitrile Heavy Duty Gloves

Product Code. 12680872 Shop All Ansell Products
Click to view available options
Size:
7
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 12680872

Brand: Ansell™ 39122070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides heavy-duty mechanical and chemical protection for wide variety of work environments. Sol-Knit™ Nitrile Heavy Duty Gloves have better mechanical performance and chemical protection.

  • Liquid and chemical protection gloves
  • Industrial gloves
  • Tough glove offers optimum comfort of use thanks to its natural shape that makes it well suited for use with long-lasting tasks
  • Superior dexterity and the high levels of wet and dry grip contribute to wearer safety and ultimately to the overall productivity
  • Rugged glove with chemical protection

Applications:

  • Adjusting panels parts components
  • Applying finish to materials, products
  • Assembly and disassembly of equipment
  • Equipment repair, maintenance
  • Handling materials with sharp or rough edges
  • Oil, fluids and filter change
  • Production line support and maintenance

Compliance

Meets ANSI A1 (cut level) and ANSI 4 (abrasion level); EN 374, EN 388, EN 407; FDA 510(k) approved.

TRUSTED_SUSTAINABILITY

Specifications

Description Anti-static, silicon-free, latex free, non-chlorinated
Applications Industrial
Coating Material Nitrile
Color Green
Latex Free Yes
Material Nitrile
Product Type Sol-Knit™ 39-122 Series Nitrile Gloves
Size 7
For Use With (Application) Heavy duty purposes
Glove Cuff Style Gauntlet
Liner Material Interlock cotton
Model 39-122 Series
Quantity 12 Pairs
Resistant To Chemical and liquid protection
Series 39-122
Length (English) 12.2 in.
Length (Metric) 310 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.