missing translation for 'onlineSavingsMsg'
Learn More

GLT25D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen GLT25D1
Concentration 0.13 mg/mL
Applications Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16836504
View Documents
Proteintech
16768-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16826504
View Documents
Proteintech
16768-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

GLT25D1
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Unconjugated
Rabbit
Rat, Mouse, Human
Q8K297, Q8NBJ5
234407, 290637, 79709
GLT25D1 Fusion Protein Ag10346
Primary
-20°C
COLGALT1
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GLT25D1
Colgalt1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications