missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glu-Glu Epitope Tag Antibody, Agarose, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Bio-Techne NB600-360
Additional Details : Weight : 0.00970kg
Description
Glu-Glu Epitope Tag Polyclonal antibody specifically detects Glu-Glu Epitope Tag in Epitope Tag samples. It is validated for Immunoprecipitation.Specifications
Glu-Glu Epitope Tag | |
Polyclonal | |
Immunoprecipitation 1:10-1:500 | |
EYMPME epitope tag, EYMPME tag, Glu-Glu epitope tag | |
Goats were immunized with a synthetic peptide representing amino acid residues 314 - 319 (EYMPME) of middle T antigen of mouse polymavirus conjugated to KLH. | |
0.1 mg | |
Epitope Tags, glu glu Epitope Tags | |
Antibody recognizes the glu-glu cleavage site. | |
IgG |
Immunoprecipitation | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4C. Do not freeze. | |
Epitope Tag |
For Research Use Only