missing translation for 'onlineSavingsMsg'
Learn More

Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

£28.34 - £144.72

Chemical Identifiers

CAS 526-95-4, 7732-18-5
Molecular Formula C6H12O7
Molecular Weight (g/mol) 196.155
MDL Number MFCD00004240
InChI Key RGHNJXZEOKUKBD-SQOUGZDYSA-N
Synonym gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
PubChem CID 10690
ChEBI CHEBI:33198
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001020
SDS View Documents
Thermo Scientific Acros
119890010
1 L Glass bottle
£28.34
1 L
Please to purchase this item. Need a web account? Register with us today!
10509610
SDS View Documents
Thermo Scientific Acros
119890025
2.5 L Glass bottle
£53.27
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10561964
SDS View Documents
Thermo Scientific Acros
119890100
10 L Plastic Drum
£144.72
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

missing translation for 'einecsNumber' : 208-401-4

missing translation for 'rtecsNumber' : LZ5057100

missing translation for 'tsca' : TSCA

Product Identifier
  • Gluconic acid, 50 wt% solution in water

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.