missing translation for 'onlineSavingsMsg'
Learn More

Gluconic acid, potassium salt, 99%, Thermo Scientific Chemicals

Product Code. 10591861
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.25
MDL Number MFCD00064211
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
Synonym potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
PubChem CID 16760467
IUPAC Name potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]

Specifications

Melting Point 178°C to 181°C
Color White
Loss on Drying 3% max. (105°C, 4 hrs) (vacuum)
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]4CO2K
Quantity 1 kg
Merck Index 15, 4492
Specific Rotation Condition +14.10° (21°C c=1.2,H2O)
Specific Rotation + 14.1
Solubility Information Solubility in water: soluble in water. Other solubilities: practically insoluble in abs. alcohol,ether,,benzene and chloroform
Formula Weight 234.25
Percent Purity 0.99
Physical Form Crystalline or Granular Powder
Chemical Name or Material Gluconic acid, potassium salt, 0.99
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'