missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gluconic acid, potassium salt, 99%, Thermo Scientific Chemicals
£57.70 - £137.00
Chemical Identifiers
Molecular Formula | C6H11KO7 |
---|---|
Molecular Weight (g/mol) | 234.25 |
MDL Number | MFCD00064211 |
InChI Key | HLCFGWHYROZGBI-JJKGCWMISA-M |
Synonym | potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal |
PubChem CID | 16760467 |
IUPAC Name | potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
SMILES | C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10318560
|
Thermo Scientific Acros
229322500 |
250 g | Plastic bottle |
£57.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10591861
|
Thermo Scientific Acros
229320010 |
1 kg | Plastic bottle |
£137.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
C6H11KO7 | |
MFCD00064211 | |
potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal | |
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
234.25 | |
HLCFGWHYROZGBI-JJKGCWMISA-M | |
16760467 | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+] |
Specifications
178°C to 181°C | |
3% max. (105°C, 4 hrs) (vacuum) | |
C6H11KO7 | |
MFCD00064211 | |
+14.10° (21°C c=1.2,H2O) | |
potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal | |
HLCFGWHYROZGBI-JJKGCWMISA-M | |
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate | |
16760467 | |
0.99 | |
Gluconic acid, potassium salt, 0.99 |
White | |
Authentic | |
HOCH2[CH(OH)]4CO2K | |
15, 4492 | |
+ 14.1 | |
Solubility in water: soluble in water. Other solubilities: practically insoluble in abs. alcohol,ether,,benzene and chloroform | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+] | |
234.25 | |
234.25 | |
Crystalline or Granular Powder |
Safety and Handling
missing translation for 'einecsNumber' : 206-074-2
missing translation for 'rtecsNumber' : LZ5230000
missing translation for 'tsca' : TSCA
RUO – Research Use Only