missing translation for 'onlineSavingsMsg'
Learn More

Gluconic acid, sodium salt, 98%

Product Code. 10604692 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
5g
This item is not returnable. View return policy

Product Code. 10604692

Brand: Thermo Scientific Acros 181390025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.137
MDL Number MFCD00064210
InChI Key UPMFZISCCZSDND-JJKGCWMISA-M
Synonym sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
PubChem CID 23672301
ChEBI CHEBI:84997
IUPAC Name sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]

Specifications

Melting Point 206.0°C to 209.0°C
Color White to Beige
pH 6.5 to 7.5 (10% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic Bottle
Linear Formula HOCH2[CH(OH)]4CO2Na
Quantity 2.5 kg
Beilstein 03, I, 188
Merck Index 15, 4492
Specific Rotation Condition + 12.10 (23.00°C c=1.75,H2O)
Specific Rotation + 12.10
Formula Weight 218.14
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Gluconic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.