missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose monohydrate, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
69.39 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Packaging | Plastic Bottle |
Quantity | 2.5 kg |
Residual Solvents | Complying |
Percent Purity | 97.5 - 102.0 % |
Grade | Reagent Pharmacopeia Organics |
Physical Form | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'