missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | GLUT1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16811395
|
Proteintech
21829-1-AP-20UL |
20 μL |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16801395
|
Proteintech
21829-1-AP-150UL |
150 μL |
£339.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
21829-1-AP antibody can also detect the 25 kDa degradation protein in SDS-PAGE.
Facilitative glucose transporter. This isoform may be responsible for constitutive or basal glucose uptake. Has a very broad substrate specificity; can transport a wide range of aldoses including both pentoses and hexoses.Specifications
GLUT1 | |
Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P11166, P17809 | |
20525, 24778, 6513 | |
GLUT1 Fusion Protein Ag16282 | |
Primary | |
-20°C | |
SLC2A1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DYT17, DYT18, GLUT, GLUT 1, GLUT1, HepG2 glucose transporter, PED, SLC2A1 | |
SLC2A1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |