missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlutaMAX™ Supplement
GlutaMAX™ Supplement
£53.25 - £590.00
Specifications
Certifications/Compliance | cGMP, ISO 13485 standard, FDA |
---|---|
Content And Storage | 15 to 30°C |
Concentration | 100 X |
Form | Liquid |
Product Type | GlutaMax |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11574466
|
Gibco
35050038 |
100 mL |
£53.25
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11584466
|
Gibco
35050087 |
20 x 100 mL |
£590.00
Pack of 20 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco™ GlutaMAX™ Supplement is an alternative to L-glutamine, with increased stability that improves cell health. GlutaMAX™ Supplement is suitable for both adherent and suspension culture of mammalian cells, with no adaptation required. GlutaMAX™ Supplement is offered as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl. GlutaMAX™ Supplement is also included in a variety of media formulations. Compared to L-glutamine, GlutaMAX™ Supplement:Features:
Minimizes toxic ammonia build-up
Unlike L-glutamine, GlutaMAX™ Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
Improves cell viability and growth
Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX™ Supplement that is not possible with L-glutamine.
Remains stable across a wide range of temperatures
GlutaMAX™ Supplement is room-temperature stable and ready to use. GlutaMAX™ Supplement retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX™ Supplement is even stable at 37°C for a week or more.
cGMP manufacturing and quality system
Gibco™ GlutaMAX™ supplement is manufactured at a cGMP compliant facility, located in Paisley, Scotland. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ GlutaMAX™ supplement made in our Grand Island, New York facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
cGMP, ISO 13485 standard, FDA | |
100 X | |
GlutaMax | |
For in-vitro diagnostic use |
15 to 30°C | |
Liquid | |
Mammalian Cell Culture |