missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ STING/TMEM173 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP2-24683PEP
Additional Details : Weight : 0.00970kg
Description
The STING/TMEM173 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
340061 | |
STING/TMEM173 | |
diH2O | |
anti-STING, endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, human STING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, mouse STING, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
M.W. (theoretical): 42 kDa | |
Cancer, Immunology, Innate Immunity |
Human | |
Store at -80 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP2-24683. For Further Blocking Peptide Related Protocol, Click Here . | |
TMEM173 | |
0.05 mg |