missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaraldehyde Solution (25%/Certified/BioReagent), Fisher BioReagents
£83.55 - £147.00
Chemical Identifiers
CAS | 111-30-8, 7732-18-5 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00007025 |
InChI Key | SXRSQZLOMIGNAQ-UHFFFAOYSA-N |
Synonym | glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde |
PubChem CID | 3485 |
ChEBI | CHEBI:64276 |
IUPAC Name | pentanedial |
SMILES | O=CCCCC=O |
Description
Chemical Identifiers
111-30-8, 7732-18-5 | |
100.12 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |
C5H8O2 | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
CHEBI:64276 | |
O=CCCCC=O |
Specifications
Liquid | |
100°C | |
1.060g/cm³ | |
24.0 to 26.5 % | |
HC(O)(CH2)3CHO | |
15, 4508 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
Certified |
-5°C | |
111-30-8, 7732-18-5 | |
16.4mmHg at 20°C | |
C5H8O2 | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
24.0 to 26.5% | |
Glutaraldehyde Solution, 25% |
Safety and Handling
DANGER!
Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1
SIGNAL WORD: DANGER!
missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : KEEP REFRIGERATED or in a very cool place to retard polymerisation
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Glutaric dialdehyde 25 % solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H317-May cause an allergic skin reaction.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Glutaraldehyde
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title