missing translation for 'onlineSavingsMsg'
Learn More
Glutaric acid, 99%, Thermo Scientific Chemicals Product Code.: 11462018

Glutaric acid, 99%, Thermo Scientific Chemicals

Product Code. 11462018
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11462018

Brand: Thermo Scientific Alfa Aesar A14595.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-94-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00004410
InChI Key JFCQEDHGNNZCLN-UHFFFAOYSA-N
Synonym glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-diShow MoreShow Less
PubChem CID 743
ChEBI CHEBI:17859
IUPAC Name pentanedioic acid
SMILES OC(=O)CCCC(O)=O

Specifications

Melting Point 95°C to 99°C
Density 1.429
Boiling Point 200°C (20 mmHg)
Quantity 100 g
Beilstein 1209725
Merck Index 14,4473
Solubility Information Soluble in water,alcohol,benzene and chloroform. Slightly soluble in petroleum ether.
Formula Weight 132.12
Percent Purity 99%
Chemical Name or Material Glutaric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253308 Certificate TypeCertificate of Analysis Date09/12/2024 Catalog NumberA14595.22,A14595.14,A14595.36
Lot NumberU29J013 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberA14595.22,A14595.14,A14595.36
Lot Number10244243 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog NumberA14595.22,A14595.14,A14595.36
Lot NumberP31I041 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14595.22,A14595.14,A14595.36
Lot Number90008825 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14595.22,A14595.14,A14595.36
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Glutaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glutaric acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.