missing translation for 'onlineSavingsMsg'
Learn More

Glutaric anhydride, 98%, Thermo Scientific Chemicals

£24.62 - £265.32

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11449073
View Documents
Thermo Scientific Alfa Aesar
A11152.18
50 g
£24.62
50g
Please to purchase this item. Need a web account? Register with us today!
11438900
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11152.22
100 g
£43.22
100g
Please to purchase this item. Need a web account? Register with us today!
11459073
View Documents
Thermo Scientific Alfa Aesar
A11152.30
250 g
£91.96
250g
Please to purchase this item. Need a web account? Register with us today!
11439073
View Documents
Thermo Scientific Alfa Aesar
A11152.0B
1000 g
£265.32
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
Specifications

Chemical Identifiers

108-55-4
114.1
VANNPISTIUFMLH-UHFFFAOYSA-N
7940
C1CC(=O)OC(=O)C1
C5H6O3
MFCD00006679
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
oxane-2,6-dione

Specifications

108-55-4
1.411 g/mL
178°C (352°F)
C5H6O3
110051
glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
VANNPISTIUFMLH-UHFFFAOYSA-N
oxane-2,6-dione
7940
98%
52°C to 57°C
291°C
98%
MFCD00006679
Moisture sensitive
Soluble in Ether,dimethyl sulfoxide and methanol and water.
C1CC(=O)OC(=O)C1
114.1
114.1
Glutaric anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H335-H315

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H318

missing translation for 'einecsNumber' : 203-593-6

missing translation for 'rtecsNumber' : MA3850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Glutaric anhydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

Supplemental information
  • MIXTURE LIST-Contains : Glutaric anhydride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only