missing translation for 'onlineSavingsMsg'
Learn More

Glutaric dialdehyde, 8% solution in water

Product Code. 10484311 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10484311

Brand: Thermo Scientific Acros 410971000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glutaric dialdehyde
Physical Form Solution
CAS 111-30-8, 7732-18-5
Density 1.0160g/mL
Packaging Glass bottle
Specific Gravity 0.918 to 1.123 (20°/20°C)
Quantity 100 g
Molecular Formula C5H8O2
Linear Formula OHC(CH2)3CHO
MDL Number MFCD00007025
Beilstein 01, 776
Fieser 01,411
Merck Index 15, 4508
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.12
Percent Purity 7 to 9 vol.%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Glutaric dialdehyde, 8% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.