missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reduced, Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Packaging:
Amber Glass
Description
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
Specifications
Specifications
Melting Point | 182°C |
Color | White |
pH | 3 |
Ignition Residue | 0.01% max. |
Loss on Drying | Report % |
Assay Percent Range | ≥98 % |
Packaging | Amber Glass |
Quantity | 1 g |
Merck Index | 15, 4511 |
Specific Rotation | -21 to -11° (+ or -) |
Show More |
Safety and Handling
Product Identifier
- Glutathione reduced
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction