missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reduced, Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Packaging:
Amber Glass
Description
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
Specifications
Specifications
Melting Point | 182°C |
Color | White |
pH | 3 |
Ignition Residue | 0.01% max. |
Loss on Drying | Report % |
Assay Percent Range | ≥98 % |
Packaging | Amber Glass |
Quantity | 25 g |
Merck Index | 15, 4511 |
Specific Rotation | -21 to -11° (+ or -) |
Show More |
Safety and Handling
Product Identifier
- Glutathione reduced
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'