missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced, Fisher BioReagents

Product Code. 10566394
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 182°C
Color White
pH 3
Ignition Residue 0.01% max.
Loss on Drying Report %
Assay Percent Range ≥98 %
Packaging Amber Glass
Quantity 100 g
Merck Index 15, 4511
Specific Rotation -21 to -11° (+ or -)
Formula Weight 307.3
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material Glutathione Reduced
Show More Show Less
Product Identifier
  • Glutathione reduced
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.