missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, 99.0 to 101.0%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Plastic bottle
Description
Used for long-term storage of bacterial cultures. Step gradients for bacteriophage/particle purification. Mammalian cell transfection. Storage buffers for enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used for long-term storage of bacterial cultures
- Step gradients for bacteriophage/particle purification
- Mammalian cell transfection
- Storage buffers for enzymes
Specifications
Melting Point | 18°C |
Density | 1.26 |
Boiling Point | 182°C (20 mmHg) |
Flash Point | 160°C |
Packaging | Plastic bottle |
Linear Formula | HOCH2CH(OH)CH2OH |
Refractive Index | 1.470-1.475 |
Quantity | 500 mL |
Beilstein | 635685 |
Specific Gravity | 1.26 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'