missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, ≥99.0% (GC), Reagent Grade, Honeywell™
Reagent Grade,≥99.0% (GC)
£46.28 - £630.59
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15617890
|
Honeywell Research Chemicals
G7757-500ML |
500 mL | Glass bottle | N/A | |||||
15697880
|
Honeywell Research Chemicals
G7757-1L |
1 L | Poly bottle |
£107.42
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15627890
|
Honeywell Research Chemicals
G7757-5L |
5 L | Aluminum bottle |
£222.14
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15607890
|
Honeywell Research Chemicals
G7757-20L |
20 L | Poly pail |
£630.59
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15687880
|
Honeywell Research Chemicals
G7757-1GA |
1 gal. | Poly bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
Colorless | |
Neutral | |
160°C (320°F) | |
Odorless | |
HOCH2CH(OH)CH2OH | |
NONH for all modes of transport | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
1mmHg (20°C) | |
Liquid |
18°C | |
1.25g/mL | |
290°C (1013 hPa) | |
698°F | |
C3H8O3 | |
MFCD00004722 | |
635685 | |
Completely miscible in water | |
OCC(O)CO | |
92.09 | |
3.1 (vs air) | |
92.09g/mol | |
≥99.0% (GC) | |
Glycerol |
Safety and Handling
missing translation for 'rtecsNumber' : 200-289-5