missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

ACS Reagent,≥99.5%

£35.68 - £1039.18

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15667890
View Documents
Honeywell-Riedel-de Haen
G7893-500ML
500 mL Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15637890
View Documents
Honeywell-Riedel-de Haen
G7893-1L
1 L Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15647890
View Documents
Honeywell-Riedel-de Haen
G7893-2L
2 L Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15658490
View Documents Promotion Details
Honeywell-Riedel-de Haen
G7893-4L
4 L Poly bottle
£323.29
4 L
Please to purchase this item. Need a web account? Register with us today!
15657890
View Documents
Honeywell-Riedel-de Haen
G7893-4X4L
4 x 4 L Glass bottle
£1039.18
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO

Specifications

56-81-5
Colorless
Neutral
160°C (320°F)
Odorless
HOCH2CH(OH)CH2OH
NONH for all modes of transport
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
CHEBI:17754
1mmHg (20°C)
Liquid
18°C
1.25g/mL
290°C (1013 hPa)
698°F
C3H8O3
MFCD00004722
635685
Completely miscible in water
OCC(O)CO
92.09
3.1 (vs air)
92.09g/mol
≥99.5%
Glycerol
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : 200-289-5

SDS
Documents

Documents

Product Certifications