missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, 99.6%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10229460

Glycerol, 99.6%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10229460
500 mL, EcoSafPak™ Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10229460

Brand: Thermo Scientific Acros 410985000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Decomposition Information >290°C
Melting Point 18.0°C
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Assay Percent Range 99.6%
Packaging EcoSafPak™ Glass Bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4716
Quantity 500 mL
Beilstein 01, 502
Merck Index 15, 4520
Specific Gravity 1.261
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Formula Weight 92.09
Vapor Pressure 0.003mbar at 50°C
Percent Purity 99.6%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464856 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number410985000,410980100,410980025
Lot NumberA0461970 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number410985000,410980100,410980025
Lot NumberA0459799 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number410985000,410980100,410980025
Lot NumberB0541563A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number410985000,410980100,410980025
Lot NumberB0541748B Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number410985000,410980100,410980025
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 99.6%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.