missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™ Product Code.: 12656707

Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 12656707
1 L, HDPE plastic bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy

Product Code. 12656707

Brand: Fisher Chemical G/0650/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
pH 5
Boiling Point 290°C
Packaging HDPE plastic bottle
Quantity 1 L
Vapor Density 3.17
Formula Weight 92.09g/mol
Vapor Pressure 0.003mbar 50
Physical Form Very Viscous Liquid
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.