missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Brand: Fisher BioReagents BP645-1
This item is not returnable.
View return policy
Description
This dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.Specifications
Glycerol Gel-Loading Dye, 5X | |
56-81-5, 7732-18-5 | |
Colorless | |
1 mL | |
Molecular Biology | |
Liquid |
Contains 30% Glycerol | |
0.25 to 0.40 AU, 0.25 to 0.45 AU at 252nm, 0.30 to 0.45 AU | |
Poly Tube | |
>1.0g/mol | |
Glycerol (30%), Proprietary Components (<6%), and Water |