Learn More
Glycerol monostearate, purified, Thermo Scientific Chemicals
£35.20 - £323.00
Chemical Identifiers
CAS | 31566-31-1 |
---|---|
Molecular Formula | C21H42O4 |
Molecular Weight (g/mol) | 358.563 |
MDL Number | MFCD00036186 |
InChI Key | VBICKXHEKHSIBG-UHFFFAOYSA-N |
Synonym | glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester |
PubChem CID | 24699 |
ChEBI | CHEBI:75555 |
IUPAC Name | 2,3-dihydroxypropyl octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304918
|
Thermo Scientific Alfa Aesar
043883.30 |
250 g |
£35.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324918
|
Thermo Scientific Alfa Aesar
043883.A1 |
1 kg |
£106.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314918
|
Thermo Scientific Alfa Aesar
043883.A7 |
5 kg |
£323.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31566-31-1 | |
358.563 | |
VBICKXHEKHSIBG-UHFFFAOYSA-N | |
24699 | |
2,3-dihydroxypropyl octadecanoate |
C21H42O4 | |
MFCD00036186 | |
glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester | |
CHEBI:75555 | |
CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O |
Specifications
31566-31-1 | |
MFCD00036186 | |
glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester | |
VBICKXHEKHSIBG-UHFFFAOYSA-N | |
2,3-dihydroxypropyl octadecanoate | |
24699 | |
358.56 | |
Glycerol monostearate, Purified |
C21H42O4 | |
14,4489 | |
Soluble in hot organic solvents;Soluble in hot water; Slightly soluble in ethanol; Insoluble in aliphatic solvents. | |
CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O | |
358.563 | |
CHEBI:75555 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 250-705-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only