missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, puriss., Honeywell

Catalog Number 15691470
Click to view available options
Quantity:
1 L
5 L
70 kg
250 kg
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
Density 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Ignition Point 698°F
Linear Formula HOCH2CH(OH)CH2OH
Quantity 250 kg
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Vapor Pressure 1mmHg (20°C)
Formula Weight 92.09g/mol
Percent Purity 99 to 101% (alkalimetric)
Grade puriss.
Chemical Name or Material Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'