missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Solution, Honeywell

Catalog Number 15691480
Click to view available options
Quantity:
1 L
5 L
25 L
70 kg
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
Density 1.252g/mL at 25°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Linear Formula HOCH2CH(OH)CH2OH · aq
Quantity 70 kg
UN Number NONH for all modes of transport
Beilstein 635685
Formula Weight 92.09g/mol (anhydrous basis)
Percent Purity 84 to 88%
Grade puriss.
Chemical Name or Material Glycerol Solution, Meets analytical specification of Ph. Eur. BP
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'