missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol triacetate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.
Solubility
Slightly miscible with water, carbon tetrachloride and carbondisulfide. Miscible with acetone, ethanol, benzene and chloroform.
Notes
Heat sensitive. Incompatible with strong oxidizing agents.
Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.
Solubility
Slightly miscible with water, carbon tetrachloride and carbondisulfide. Miscible with acetone, ethanol, benzene and chloroform.
Notes
Heat sensitive. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 2°C to 4°C |
Density | 1.155 |
Boiling Point | 257°C to 259°C |
Flash Point | 149°C (300°F) |
Assay Percent Range | 99% |
Odor | Petroleum-like |
Quantity | 2500 g |
Solubility Information | Slightly miscible with water,carbon tetrachloride and carbondisulfide. Miscible with acetone,ethanol,benzene and chloroform. |
Formula Weight | 218.21 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'