Learn More
Glycerol tributyrate, 97%, Thermo Scientific Chemicals
Glycerol tributyrate, 98%, C15H26O6, CAS Number-60-01-5, tri-n-butyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, glycerol tributyrate, glycerol tributanoate, glyceroltributyrin, tributyrin, butyrin, butyryl triglyceride, tributin, 1000g, 1714746, 307 deg.C to 308 deg.C, CHEBI:35020
£20.50 - £156.78
Chemical Identifiers
CAS | 60-01-5 |
---|---|
Molecular Formula | C15H26O6 |
Molecular Weight (g/mol) | 302.367 |
MDL Number | MFCD00009392 |
InChI Key | UYXTWWCETRIEDR-UHFFFAOYSA-N |
Synonym | tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin |
PubChem CID | 6050 |
ChEBI | CHEBI:35020 |
IUPAC Name | 2,3-di(butanoyloxy)propyl butanoate |
SMILES | CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433784
|
Thermo Scientific Alfa Aesar
A11830.18 |
50 g |
£20.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443784
|
Thermo Scientific Alfa Aesar
A11830.30 |
250 g |
£45.73
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423784
|
Thermo Scientific Alfa Aesar
A11830.0B |
1000 g |
£156.78
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents, Strong acids, Strong bases. Use with adequate ventilation.
Chemical Identifiers
60-01-5 | |
302.367 | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
6050 | |
2,3-di(butanoyloxy)propyl butanoate |
C15H26O6 | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CHEBI:35020 | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Specifications
60-01-5 | |
1.032 | |
174°C (345°F) | |
1.4358 | |
1714746 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
2,3-di(butanoyloxy)propyl butanoate | |
6050 | |
302.37 | |
Glycerol tributyrate |
-75°C | |
307°C to 308°C | |
C15H26O6 | |
MFCD00009392 | |
14,9620 | |
Not miscible in water. | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
302.367 | |
CHEBI:35020 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 200-451-5
missing translation for 'rtecsNumber' : ET7350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.