missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.
Solubility
Miscible with benzene, alcohol and ether. Slightly miscible with water.
Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases and peroxides.
Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.
Solubility
Miscible with benzene, alcohol and ether. Slightly miscible with water.
Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases and peroxides.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -82°C |
Density | 1.078 |
Boiling Point | 189°C |
Flash Point | 83°C (181°F) |
Refractive Index | 1.45 |
Quantity | 250 g |
UN Number | UN2810 |
Beilstein | 2506 |
Solubility Information | Miscible with benzene,alcohol and ether. Slightly miscible with water. |
Formula Weight | 142.16 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'