missing translation for 'onlineSavingsMsg'
Learn More

Glycine, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10407873
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg

Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Glycine is an essential acid that can be used for the synthesis of creatine, purines, and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity


Applications

  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 233°C
Arsenic (As) 3ppm max.
pH 5.5 to 6.5 (5% Aqueous Solution)
Color Colorless to Yellow
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 0.98
Infrared Spectrum Authentic
Linear Formula H2NCH2CO2H
Beilstein 04, 333
Fieser 1412
Loss on Drying 0.2% max. (105°C, 2 hrs)
Merck Index 15, 4526
Packaging Plastic Bucket
Residue after Ignition 0.1% max.
Formula Weight 75.07
Percent Purity ≥97.5% (Total Base)
Grade Extra Pure
Density 1.5950g/mL
Quantity 25 kg
Physical Form Liquid
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'