missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, 98%, extra pure, Thermo Scientific Chemicals
Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins.
£27.50 - £647.00
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10784611
|
Thermo Scientific Acros
120072500 |
250 g |
£27.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10408460
|
Thermo Scientific Acros
120070010 |
1 kg |
£66.90
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10637652
|
Thermo Scientific Acros
120070050 |
5 kg |
£203.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10407873
|
Thermo Scientific Acros
120070250 |
25 kg |
£647.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Glycine is an essential acid that can be used for the synthesis of creatine, purines, and porphyrins
- Soluble in water; has a pH of ∼=5.5-6.5 in solution
- Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity
Applications
- Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
- Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
- Intended for biochemistry applications
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Specifications
233°C | |
3ppm max. | |
Colorless to Yellow | |
0.98 | |
Authentic | |
MFCD00008131 | |
1412 | |
15, 4526 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
NCC(O)=O | |
0.1% max. | |
750 | |
75.07 | |
Extra Pure | |
Liquid |
56-40-6 | |
5.5 to 6.5 (5% Aqueous Solution) | |
20ppm max. | |
C2H5NO2 | |
H2NCH2CO2H | |
04, 333 | |
0.2% max. (105°C, 2 hrs) | |
Plastic Bottle | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
75.07 | |
CHEBI:15428 | |
≥97.5% (Total Base) | |
1.5950g/mL | |
Glycine |
RUO – Research Use Only