missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Glycine, 99% Product Code.: 11454894

Thermo Scientific Chemicals Glycine, 99%

Product Code. 11454894
500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11454894

Brand: Thermo Scientific Chemicals A13816.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine is used as an additive in pet food, animal feed and sweetener and taste enhancer in human foods. It is also used as a buffering agent in antacids, analgesics, antiperspirants, cosmetics and toiletries. It acts as a neurotransmitter in the central nervous system, especially in the spinal cord, brainstem, and retina. It serves as a buffer agent and prevents the sample damage during electrophoresis. Further, it is used to remove protein-labelling antibodies from western blot membranes. In addition to this, it is involved in the production of rubber sponge products, metal complexants and fertilizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is used as an additive in pet food, animal feed and sweetener and taste enhancer in human foods. It is also used as a buffering agent in antacids, analgesics, antiperspirants, cosmetics and toiletries. It acts as a neurotransmitter in the central nervous system, especially in the spinal cord, brainstem, and retina. It serves as a buffer agent and prevents the sample damage during electrophoresis. Further, it is used to remove protein-labelling antibodies from western blot membranes. In addition to this, it is involved in the production of rubber sponge products, metal complexants and fertilizers.

Solubility
Soluble in water and pyridine. Slightly soluble in acetone. Insoluble in diethyl ether, n-octanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
pH 5.97
Assay Percent Range 99%
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in water and pyridine. Slightly soluble in acetone. Insoluble in diethyl ether,n-octanol and ethanol.
Density 1.1607
Quantity 500 g
Odor Odorless
Formula Weight 75.07
Percent Purity 99%
Chemical Name or Material Glycine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY25K020 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog NumberA13816.0C,A13816.36,A13816.0E
Lot NumberX21K007 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberA13816.0C,A13816.36,A13816.0E
Lot NumberW26K021 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberA13816.0C,A13816.36,A13816.0E
Lot NumberU28K024 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog NumberA13816.0C,A13816.36,A13816.0E
Lot NumberA0464721 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog NumberA13816.0C,A13816.36,A13816.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Glycine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.