missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Glycine, Electrophoresis Grade, 99+% Product Code.: 15406349

Thermo Scientific Chemicals Glycine, Electrophoresis Grade, 99+%

Product Code. 15406349
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 15406349

Brand: Thermo Scientific Chemicals J64365.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin?s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin′s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
pH 5.97
Color White
Linear Formula H2NCH2CO2H
Beilstein 635782
Merck Index 14,4491
Solubility Information Freely soluble in water
Density 1.595
Quantity 1 kg
Odor Odorless
Formula Weight 75.07
Percent Purity ≥99.5%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Glycine, Electrophoresis Grade, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.