Learn More
Thermo Scientific Chemicals Glycine hydrochloride, 98+%
£44.12 - £493.46
Chemical Identifiers
CAS | 6000-43-7 |
---|---|
Molecular Formula | C2H6ClNO2 |
Molecular Weight (g/mol) | 111.53 |
MDL Number | MFCD00012872 |
InChI Key | IVLXQGJVBGMLRR-UHFFFAOYSA-N |
Synonym | glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride |
PubChem CID | 22316 |
IUPAC Name | 2-aminoacetic acid;hydrochloride |
SMILES | [H+].[Cl-].NCC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10156800
|
Thermo Scientific Chemicals
411011000 |
100 g | Glass Bottle |
£44.12
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10279790
|
Thermo Scientific Chemicals
411015000 |
500 g | Glass Bottle |
£137.69
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10013733
|
Thermo Scientific Chemicals
411010025 |
2.5 kg | Plastic Bottle |
£493.46
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6000-43-7 | |
111.53 | |
IVLXQGJVBGMLRR-UHFFFAOYSA-N | |
22316 | |
[H+].[Cl-].NCC(O)=O |
C2H6ClNO2 | |
MFCD00012872 | |
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride | |
2-aminoacetic acid;hydrochloride |
Specifications
178.0°C to 182.0°C | |
1 to 2 (10% aq. soln.) | |
Authentic | |
C2H6ClNO2 | |
MFCD00012872 | |
15, 4526 | |
Solubility in water: soluble. | |
IVLXQGJVBGMLRR-UHFFFAOYSA-N | |
2-aminoacetic acid;hydrochloride | |
22316 | |
≥98% | |
Glycine hydrochloride, 99% |
6000-43-7 | |
White | |
99% | |
NH2CH2COOH·HCl | |
04, 340 | |
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride | |
1% Max. (K.F.) | |
[H+].[Cl-].NCC(O)=O | |
111.53 | |
111.53 | |
Crystalline Powder |
Safety and Handling
MOISTURE SENSITIVE
missing translation for 'einecsNumber' : 227-841-8
missing translation for 'rtecsNumber' : MC0560000
missing translation for 'tsca' : TSCA
- Glycine hydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.