missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Click to view available options
Quantity:
1 kg
5 kg
Description
Protein Electrophoresis & Blotting
Specifications
Assay Percent Range | ≥99% |
Formula Weight | 75.07 |
Grade | Molecular Biology |
Density | 1.16g/mL (Lit.) |
Quantity | 5 kg |
Physical Form | Powder |
Chemical Name or Material | Glycine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'