missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 500 g |
Formula Weight | 75.07 |
Specific Gravity | 1L = 1.16kg |
Grade | Multi-Compendial/USP |
Chemical Name or Material | Glycine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glycine, Multi-Compendial, U.S.P., J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction