missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-butyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7046765
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6456-74-2
C6H13NO2
131.175
MFCD00038194
SJMDMGHPMLKLHQ-UHFFFAOYSA-N
tert-butyl glycinate, glycine tert-butyl ester, glycine, 1,1-dimethylethyl ester, tert-butyl aminoacetate, h-gly-otbu, t-butyl glycinate, glycin-tert-butyl ester, 1,1-dimethylethyl glycinate, glycine t-butyl ester, aminoacetic acid tert-butyl ester
151417
tert-butyl 2-aminoacetate
CC(C)(C)OC(=O)CN
This item is not returnable. View return policy
6456-74-2
C6H13NO2
131.175
MFCD00038194
SJMDMGHPMLKLHQ-UHFFFAOYSA-N
tert-butyl glycinate, glycine tert-butyl ester, glycine, 1,1-dimethylethyl ester, tert-butyl aminoacetate, h-gly-otbu, t-butyl glycinate, glycin-tert-butyl ester, 1,1-dimethylethyl glycinate, glycine t-butyl ester, aminoacetic acid tert-butyl ester
151417
tert-butyl 2-aminoacetate
CC(C)(C)OC(=O)CN

Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

Solubility
Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 29°C to 31°C (2mmHg)
Beilstein 2039000
Refractive Index 1.424
Solubility Information Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.
Formula Weight 131.18
Percent Purity 97%
Density 0.96
Quantity 1 g
Sensitivity Air sensitive
Chemical Name or Material Glycine tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'