missing translation for 'onlineSavingsMsg'
Learn More

Glycine, U.S.P. - F.C.C., J.T. Baker™

Product Code. 13215403
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Formula Weight 75.07
Grade FCC/USP
Quantity 500 g
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'