missing translation for 'onlineSavingsMsg'
Learn More

Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™

Product Code. 10754724
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10754724

Brand: Fisher BioReagents BP3815

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Glycine (White Crystals or Crystalline Powder)

Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 182°C
Absorbance 0.1 AU max. at 280nm
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 5.9 to 6.4
Color White
Chloride 0.003% max.
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.10% max.
Identification Pass Test
Iron (Fe) 7ppm max.
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.20% max.
Other Amino Acids Pass Test
Packaging Poly Pail
Merck Index 15, 4526
Density 1.595g/cm³
Quantity 5 kg
Formula Weight 75.07
Percent Purity 98.5 to 101.0%
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.